3,4-Dihydroxybenzylamine hydrobromide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-(Aminomethyl)benzene-1,2-diol hydrobromide
CAS Number:16290-26-9
:
Formula:C₇H₁₀BrNO₂
MDL Number:MFCD00012859
Molecular Weight:220.07 g/mol
Melting Pt:>180 °C
Storage Temperature:Ambient
EINECS:240-382-8
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light beige Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together