Order Entry
Slovenia
ContactUsLinkComponent
Kjeldhal steam distillation units
 :  BEHR
undefined
Kjeldhal steam distillation units
 :  BEHR
  • Catalog No:
  • BEHRB00218034
  • 705-0425
  • 705-0426
  • 705-0423
  • 705-0427
  • 705-0424
  • BEHRB00233702
  • Model:
  • S 5
  • S 4
  • S 1
  • S 3
  • S 2
  • Water consumption:
  • 300 l/h
  • 300 l/h
  • 300 l/h
  • 300 l/h
  • 300 l/h
  • Programs:
  • 99
  • 99
  • 1
  • 10
  • 1
  • Weight:
  • 29 kg
  • 24,5 kg
  • 20 kg
  • 23 kg
  • 21 kg
  • W×H×D:
  • 410×675×410 mm
  • 410×675×410 mm
  • 410×675×410 mm
  • 410×675×410 mm
  • 410×675×410 mm
Frequently Bought Together