Capillary columns, OPTIMA® δ
Medium polar GC phases with autoselectivity.
- Analytes determine the polarity of the phase
- Unique from MN, no similar phase
- Ideal for MSD and PND detectors
For columns with 0,10 to 0,32 mm ID the max. temperature for isothermal operation is 340 °C, the max. temperature for short isotherms in a temperature program is 360 °C; for 0,53 mm ID columns the max. temperatures are 320 and 340 °C, respectively.
Polysiloxane phase without CN groups (better bleeding behaviour).
OPTIMA® δ-3: Autoselective polarity between polarities 5 and 1701.
OPTIMA® δ-6: Autoselective polarity between polarities 17 and 210.