7-Methoxybenzofuran-2-carboxylic acid ≥97%
Catalog # ALFAB21185.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:192.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:4790-79-8
Formula:C₁₀H₈O₄
MDL Number:MFCD00060516
Melting Pt:217…220 °C