4-(Trifluoromethyl)cyclohexanone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75091-99-5
:
Formula:C₇H₉F₃O
MDL Number:MFCD00102145
Boiling Pt:67…68 °C (16 mmHg)
Molecular Weight:166.14 g/mol
UN:1224
ADR:3 III
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: