Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4,5-Dimethoxy-2-nitrophenyl)acetic acid
(4,5-Dimethoxy-2-nitrophenyl)acetic acid ≥97%
(4,5-Dimethoxy-2-nitrophenyl)acetic acid ≥97%
:
Thermo Fisher Scientific
(4,5-Dimethoxy-2-nitrophenyl)acetic acid ≥97%
:
Thermo Fisher Scientific
:
2-(4,5-Dimethoxy-2-nitrophenyl)acetic acid
:
73357-18-3
:
:
C₁₀H₁₁NO₆
:
MFCD00118466
:
241.20 g/mol
:
206…208 °C
:
GHS07
:
Ambient
Environmentally Preferable:
msds_ALFAA18884
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years