(R)-(+)-Indoline-2-carboxylic acid ≥97%
Catalog # ALFAH27626.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:163.18 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:98167-06-7
Formula:C₉H₉NO₂
MDL Number:MFCD00792496
Melting Pt:∼ 177 °C (dec.)