Glycylglycine (Gly-Gly)
Buffer with a useful pH range 7,5 to 8,9
- EINECS: 209-127-8
- GHS symbol and signal word: GHS07 and warning
- Melting point: ca 220 °C (dec.)
- Merck index: 14,4503
Purity: 99% CAS No.: 556-50-3 Molecular Formula: H2NCH2CONHCH2CO2H Formula Weight: 132.12 Melting Point: ca 220° dec. Beilstein Reference: 1765223 Merck Reference: 14,4503 MDL No.: MFCD00008130
- Source:
- Species:
- Environmentally Preferable: