tetra-Chloroauric(III) acid (40 - 44% Au) (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Auric chloride, tetra-Chloroauric acid, Gold chloride, Gold trichloride, Hydrogen tetrachloroaurate (III)
CAS Number:16903-35-8
:
Formula:HAuCl₄
MDL Number:MFCD00011322
Density:3.9
Molecular Weight:339.78 g/mol
ADR:8 III
Merck Index:13,04536
EINECS:240-948-4
UN:3264
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: