Safety spectacles, uvex meteor
Catalog # UVEX9134.005
Supplier: UVEX
Specifications
- Type:Anti-UV (2-1,2), anti-scratch, chemical resistant
- Colour:Transparent
- Seq:02
- Environmentally Preferable:
- Lenses:Clear PC, Optidur NC
- Pk:1
Specifications
About this item
The dual lens glasses with close fitting frame provide reliable protection and protection against particles from below.
- Anatomically shaped nose bridge
- Side shields and upper eye rim protection integrated in frame design
- Side arms individually adjustable in four different lengths
- Adjustable comfortable soft-form ear pieces
Certifications: EN 166
Delivery: Supplied with case and microfibre cloth.