Order Entry
Singapore
ContactUsLinkComponent
Acetonitrile for HPLC, for spectrophotometry, J.T.Baker®
Acetonitrile for HPLC, for spectrophotometry, J.T.Baker®
Catalog # BAKR9152-80
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Acetonitrile for HPLC, for spectrophotometry, J.T.Baker®
Catalog # BAKR9152-80
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:
    ≤0.005
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤190
  • Gradient Elution Test (a.u.) - 254 nm:
    ≤0.005
  • Gradient Elution Test (a.u.) - 210 nm:
    ≤0.005
  • Density (g/mL) at 25°C:
    0.775 - 0.780
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.3 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay (CH₃CN) (by GC):
    ≥99.9 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Titrable Acid (µeq/g):
    ≤0.8
  • Titrable Base (µeq/g):
    ≤0.6
  • Water (by KF, coulometric):
    ≤100 ppm
  • Carbonyl Compounds (as Acetone):
    ≤25.0 ppb

Specification Test Results

About this item

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
 :
 :41.05 g/mol
Flash Pt:2 °C (closed cup)
EINECS:200-835-2
UN:1648
REACH:01-2119471307-38
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,00071