Order Entry
Singapore
ContactUsLinkComponent
Biohazard spill kit
Biohazard spill kit
Catalog # GUESH8616
Supplier:  GUEST
undefined
Biohazard spill kit
Catalog # GUESH8616
Supplier:  GUEST
Specifications
  • Description:
    Biohazard spill kit
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The biohazard spill kit contains everything needed to clean up spills of blood or other biohazardous fluid quickly and safely. Each kit contains.
  • Haz-Tab granules, which are formulated using sodium dichloroisocyanurate, a powerful chlorinating agent effective against bacteria, fungi and viruses; poured onto the spill they absorb and disinfect simultaneously
  • A scoop and scraper to transfer the spill, including broken glass, to the yellow incineration bag for disposal
  • Tablets to enable preparation of a 10000 ppm solution of chlorinating agent, with which splashes can be wiped up
Delivery: Kit contains: 2 tubs Haz-Tab granules, 1 Haz-Tab diluter, 1 tub Haz-Tab tablets, 100 disposable gloves, 20 scoops and scrapers, 20 paper towels, 25 disposal bags, 3 aprons and full instructions.