Bis(phenylthio)methane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3561-67-9
:
Formula:C₁₃H₁₂S₂
MDL Number:MFCD00003067
Boiling Pt:194 °C (8 mmHg)
Molecular Weight:232.37 g/mol
Melting Pt:36…38 °C
Storage Temperature:Ambient
EINECS:222-624-4
Specifications
- Environmentally Preferable: