Manual cell counter, Tally
Catalog # HEATHS6594
Supplier: HEATHROW
Specifications
- Range:0 − 9999
- Description:Manual cell counter, tally
- Material:Chrome plated
- Seq:01
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This counter is suitable for counting blood cells, bacterial colonies, drops of liquid.
- Quick-reset knob
- Finger ring
Wר: 41×46 mm.