Counting chambers, Imp. Neubauer
Supplier: MARIENFELD
Certificates
About this item
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
Certifications: IVD directive 98/79/EC.
Delivery: Supplied with two cover glasses.