Order Entry
Singapore
ContactUsLinkComponent
 
Sodium fluoride ≥98%, purified
Catalog # VWRC27858.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium fluoride ≥98%, purified
Catalog # VWRC27858.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :41.99 g/mol
Boiling Pt:1700
EINECS:231-667-8
UN:1690
 :GHS06
Storage Temperature:Ambient
CAS Number:7681-49-4
Formula:NaF
MDL Number:MFCD00003524
Melting Pt:993
Density:2.8 g/cm³ (20 °C)
ADR:6.1 III
Merck Index:13,08691