Order Entry
Singapore
ContactUsLinkComponent
Sodium carbonate ≥98%, purified
Sodium carbonate ≥98%, purified
Catalog # VWRC27762.367
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium carbonate ≥98%, purified
Catalog # VWRC27762.367
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

Synonyms: Calcined soda, Carbonic acid disodium salt
 :
 :105.99 g/mol
Boiling Pt:1600
EINECS:207-838-8
REACH:01-2119485498-19
 :GHS07
Storage Temperature:Ambient
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494
Melting Pt:854
Density:2.53 g/cm³ (20 °C)
Merck Index:14,08596