Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Bromoethyl)cyclohexane
(2-Bromoethyl)cyclohexane ≥99%
(2-Bromoethyl)cyclohexane ≥99%
:
Thermo Fisher Scientific
(2-Bromoethyl)cyclohexane ≥99%
:
Thermo Fisher Scientific
:
2-Cyclohexylethyl bromide, 1-Bromo-2-cyclohexylethane
:
1647-26-3
:
:
C₈H₁₅Br
:
MFCD00019398
:
70…71 °C (6 mmHg)
:
1.221
:
191.11 g/mol
:
96 °C (204 °F)
:
Ambient
:
216-712-1
:
GHS07
Environmentally Preferable:
msds_ALFAL09466
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years