Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥97%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥97%
:
Thermo Fisher Scientific
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥97%
:
Thermo Fisher Scientific
:
1-Nitro-3,5-bis(trifluoromethyl)benzene
:
328-75-6
:
:
C₈H₃F₆NO₂
:
MFCD00000384
:
75…76 °C (11 mmHg)
:
1.535
:
259.11 g/mol
:
77 °C (170 °F)
:
Ambient
:
206-336-6
:
GHS07
Environmentally Preferable:
msds_ALFAA16797
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years