Tubing, PTFE
: KARTELL
PTFE, white.
- Excellent chemical resistance
- Very good thermal resistance, autoclavable
- Free of extractable substances, physiologically neutral, biocompatible
Excellent resistance to acids, bases and solvents.
Temperature resistance: −200 to +260 °C