Bottles, wide neck, PTFE, with screw caps, PTFE
Supplier: KARTELL
Certificates
About this item
PTFE, white.
- Very good mechanical and chemical resistance
- Can be safely used continuously from −270° C up to 250 °C
- Suitable for foodstuff
Particularly suitable for dissolutions and digestions with acids and corrosive agents. Entire bottle, including screw caps and inserts made of PTFE.