Bottles, aluminium, with screw cap
Supplier: BURKLE
Certificates
About this item
Pure aluminium (material AL 99,5). This aluminium bottle is particularly suitable for storing, transport, or sending substances and reference samples which have to be stored completely unaltered.
- With tamper-evident screw cap made of PP with aluminium inner seal
- Suitable for food (exception: highly acidic and salt-containing foods)
- With UN approval (for 60 to 5600 ml)
- Can be completely emptied without residues
- Odour-neutral
- Impervious to light, rustproof