4-Bromo-3-nitroanisole 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Bromo-4-methoxy-2-nitrobenzene
CAS Number:5344-78-5
:
Formula:C₇H₆BrNO₃
MDL Number:MFCD00051511
Boiling Pt:153…154 °C (13 mmHg)
Molecular Weight:232.03 g/mol
Melting Pt:29…32 °C
Flash Pt:>110 °C (230 °F)
EINECS:226-290-0
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: