9,10-Dimethylanthracene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:781-43-1
:
Formula:C₁₆H₁₄
MDL Number:MFCD00001262
Boiling Pt:371
Molecular Weight:206.29 g/mol
Melting Pt:182…184 °C
EINECS:212-308-4
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: