Citric acid, anhydrous 99.5-100.5%, powder Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
Boiling Pt:310
Melting Pt:149…151 °C
Density:1.665 g/cm³ (18 °C. OECD 109)
Molecular Weight:192.13 g/mol
Merck Index:13,02350
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (calculated on anhydrous):99.5 - 100.5 %
- Appearance:Conforms (see CoA/CoS)
- Identification B:Passes test
- Appearance of solution:Passes test
- Readily carbonisable substances:Passes test
- Oxalic acid:Max. 360 ppm
- SO4 (Sulphate):Max. 150 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Water:Max. 1.0 %
- Sulphated ash:Max. 0.1 %
- Residual solvents:Unlikely by manuf.process
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 50L Plastic bucket UN Y (ZPACK13247050)
Packaging Information
20282_CITRIC ACID POWDER PH.EUR_DIOXIN_REV0 20282_CITRIC ACID POWDER PH.EUR_MELAMINE_REV0 20282_CITRIC ACID POWDER PH.EUR_NANO_REV0 Citric Acid, Anhydrous 99,5 - 100,5%, Powder Ph. Eur.
Additional Product Details
spec_VWRC20282.293 spec_VWRC20282.293 spec_VWRC20282.362 spec_VWRC20282.362 spec_VWRC20282.362
Product specification
More
VWR Chemicals for Biopharma and Pharma Production