Aspirating pipette, Corning®
Catalog # CORN4930
Supplier: Corning
Specifications
- Description:Vacuum aspirator
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This aspirating device is used for the removal of liquids from a variety of lab vessels. It is designed for use with an in-house vacuum system. The aspirator allows precise fingertip control of aspiration and presents a sensible alternative to the use of glass pasteur pipettes for supernatant disposal.
- Designed for a comfortable fit in either hand
- Can be used with a variety of collection devices
Designed for use with a vacuum pump or an in-house vacuum system and a suitable collection vessel
Delivery: Supplied with handpiece, grommet for accessory attachment and single-channel adapter for disposable pipette tips (1 to 200 µl). An 8-channel adapter is available separately.