1-Bromohexane ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Hexyl bromide
CAS Number:111-25-1
:
Formula:BrH₂C(CH₂)₄CH₃
MDL Number:MFCD00000271
Boiling Pt:156 °C
Melting Pt:–85 °C
Density:1.176
Molecular Weight:165.07 g/mol
Flash Pt:50 °C (122 °F)
Storage Temperature:Ambient
EINECS:203-850-2
UN:1993
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable: