Sodium carbonate decahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Soda
CAS Number:6132-02-1
:
Formula:Na₂CO₃·10H₂O
MDL Number:MFCD00149178
Density:1.46
Molecular Weight:286.14 g/mol
Melting Pt:32…34 °C
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: