Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.004 g/cm³ (25 °C)
Molecular Weight:36.46
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0998 - 0.1002 mol/l
- Conforms to Reag. Ph.Eur.:Passes test
- NIST traceable:Confirmed