Hydrochloric acid 37% Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.18 g/cm³ (20 °C)
Molecular Weight:36.46
Melting Pt:–30 °C
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8 II
REACH:01-2119484862-27
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:35.0 - 39.0 %
- Appearance:Clear colourless liquid
- Identification A:Passes test
- Identification B:Passes test
- Identification C:Passes test
- Appearance of solution:Passes test
- Free chlorine:Max. 4 ppm
- SO4 (Sulphate):Max. 20 ppm
- Heavy metals (as Pb):Max. 2 ppm
- Residue on evaporation:Max. 0.01 %
- Residual solvents:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250) 25L Plastic drum for liquids DIN 61 (ZPACK13952503)
Packaging Information
20255_HYDROCHLORIC ACID 37% PH.EUR_BSE_REV1
BSE/TSE Certificates
20255_HYDROCHLORIC ACID 37% PH.EUR_DIOXIN Hydrochloric Acid 37% Ph. Eur.
Additional Product Details
spec_VWRC20255.290 spec_VWRC20255.290 spec_VWRC20255.324 spec_VWRC20255.324 spec_VWRC20255.368
Product specification
More