Order Entry
Singapore
ContactUsLinkComponent
Safety drying oven, FDL 115
Safety drying oven, FDL 115
Catalog # BIND9010-0292
Supplier:  Binder
undefined
Safety drying oven, FDL 115
Catalog # BIND9010-0292
Supplier:  Binder

Specifications

  • Capacity:
    115 L
  • Convection type:
    Forced convection
  • Temperature range:
    Ambient +5…300 °C
  • Description:
    FDL 115
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    FDL 115
  • Temperature variation (spatial):
    ±1,5 °C @ 70 °C
    ±2,5 °C @ 150 °C
    ±4 °C @ 300 °C
  • Ext. W×D×H:
    835×685×800 mm
  • Int. W×D×H:
    600×435×435 mm
  • Max. load per shelf:
    20 kg
  • Power consumption:
    2900 W
  • Shelves supplied (max.):
    2 (5)
  • Weight:
    90 kg
  • Temperature fluctuation (time):
    @ 150 °C: ±0,3 °C
  • Pk:
    1

Specifications

About this item

These drying ovens for limited quantities of solvents, have high temperature accuracy and silicone- and dust-free stainless steel chambers. Units are widely used for applications such as quality assurance of paints or other liquid coatings. FDL ovens feature an MP controller with adjustable ramp function and integrated weekly program timer with real time function.

  • APT.line™ pre-heating chamber technology with symmetrical airflow and defined velocities for reproducible drying
  • Fulfils all safety regulations for handling materials containing solvents in accordance with EN 1539
  • Digital temperature setting
  • Calibration and validation possible

FDL 115 has an adjustable temperature safety device, Class 2 (DIN 12880) with visual and acoustic alarm, and rear 100 mm Ø exhaust duct. Units have a replaceable Class F6 (for fine particle sizes between 1 and 10 µm) fresh air filter cartridge and fresh air monitoring with visual and acoustic alarm and automatic heating shut off. GLP/GMP compliance is assisted by the RS422 interface and optional APT-COM™ DataControlSystem software.

Highest permitted solvent quantity: 6,65 / 0,014 (g/lbs.) (at T-180 °C, M-100 g/mol, U-40 g/m³, K=0,5) (g), T = drying temperature, M = molecular mass, U = lower explosion limit and K = solvent vapour concentration as percentage of lower explosion limit.

Certifications: IP protection class according to DIN EN 60529: IP 33.
Delivery: Supplied with two chrome plated shelves.