Digital barometer, 511
TEST0560.0511
: Testo
- Range:300 - 1200 hPa
- Resolution:0,1 hPa
- Weight:90 g
- W×D×H:119×46×25 mm
- Environmentally Preferable:
- Accuracy:±3,0 hPa
- Pk:1
Pocket absolute pressure meter. Very handy, small and easy to use, this barometer permits measurements of barometric air pressure by entering on-site height above sea level and barometric elevation measurement between two points. Backlit display completes the instrument.
- Automatic off function
- Eight switchable pressure units: hPa, mbar, Pa, mmH₂O, mmHg, inH₂O, inHg, psi
- Barometric altitude measurement
: Supplied with protective cap, wrist strap, belt holder and two alkaline AAA batteries for about 500 hours of use without display illumination.