Tubing, Tygon® A-60-G
New Product
Thermoplastic elastomer, black.
- Long lasting, weatherproof, good abrasion resistance and exceptional resistance to fatigue caused by continual flexing
- Excellent chemical resistance
- Low gas permeability compared with rubber tubing
Versatile tubing for industrial and laboratory applications. Heat- and ozone-resistant. Excellent resistance to inorganic liquids (acids and bases). Ideal for use in peristaltic pumps.
Temperature resistance: −60 to +135 °C
Hardness (Shore durometer A): 61
Density: 0,98