Spore strips, Sportrol™
About this item
Biological sterilisation indicators are used to perform sterilisation cycle validation. They are created by inoculating spore suspensions of certified population and resistance onto paper strips.
- Standard spore strips and combination spore strips are 7×38 mm
- Mini spore strips are 2×10 mm in size and will fit into small areas of a device where a standard sized spore strip cannot be used, such as within a syringe barrel, inside tubing or under a cap
- All spore strips have a shelf life of 24 months when stored under labelled conditions, except Geobacillus stearothermophilus strips 489-0081 and 489-0096 (18 months)
Meet USP and ISO 11128-1 standards.
Certifications: Meet USP and ISO 11138-1 standards.
Ordering information: Spore strips are individually packaged in glassine envelopes in packs of 100. Supplied with instructions for use, certified population, D-value, Z-value, survival/kill data, and expiry date.
4 Options Available Below
Product Details & Documents
Biological sterilisation indicators are used to perform sterilisation cycle validation. They are created by inoculating spore suspensions of certified population and resistance onto paper strips.
- Standard spore strips and combination spore strips are 7×38 mm
- Mini spore strips are 2×10 mm in size and will fit into small areas of a device where a standard sized spore strip cannot be used, such as within a syringe barrel, inside tubing or under a cap
- All spore strips have a shelf life of 24 months when stored under labelled conditions, except Geobacillus stearothermophilus strips 489-0081 and 489-0096 (18 months)
Meet USP and ISO 11128-1 standards.
Certifications: Meet USP and ISO 11138-1 standards.
Ordering information: Spore strips are individually packaged in glassine envelopes in packs of 100. Supplied with instructions for use, certified population, D-value, Z-value, survival/kill data, and expiry date.
Documents
Product Family Options
Product Information
- AvailabilityPrice
- NAMSBG-106Standard, Bacillus atrophaeus 10⁶ spores/strip, ethylene oxide
- NAMSBS-106Standard, Geobacillus stearothermophilus, 10⁶ spores/strip, steam sterilisationStandard, Geobacillus stearothermophilus, 10⁶ spores/strip, steam sterilisationLog in for availability1 * 100 items
- NAMSSTN-062Mini, Bacillus atrophaeus 10⁶ spores/strip, ethylene oxide or dry heat sterilisationMini, Bacillus atrophaeus 10⁶ spores/strip, ethylene oxide or dry heat sterilisationLog in for availability1 * 100 items
- NAMSSTS-062Mini, Geobacillus stearothermophilus, 10⁶ spores/strip, steam sterilisationMini, Geobacillus stearothermophilus, 10⁶ spores/strip, steam sterilisationLog in for availability1 * 100 items