Chamber slides, Nunc™ Lab-Tek™ II
PS medium chamber on glass slide. PS lid.
- Removable medium chamber of 1, 2, 4 and 8-well configuration, attached to non-fluorescent glass microscope slide
- Rounded corners using biocompatible adhesive
- Inert hydrophobic well border printed on slide
- Superfrost™ printed writing area
- Treated to ensure excellent cell attachment and growth
Designed for cell culture, viral and mycoplasma testing, chromosome studies, toxic tests and immunocytology.
- Catalog No:
- NUNC154453
- NUNC154526
- NUNC154461
- NUNC154534
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement