Anion exchange HPLC columns, PRP-X100
Supplier: HAMILTON BONADUZ
Certificates
About this item
The PRP-X100 is a polymeric column packing for separation of inorganic and organic anions from 10 to 500 ppm. These columns can be used to separate ions at concentrations from 1 to 1000 ppm without suppression or elaborate preconcentration techniques. The highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0 - 100% aqueous or organic; pH 1 to 13), effectively lengthening column life.
- Easily separate difficult anions such as cyanide, borate and silicate at high pH (11,5)
These columns can be used with any existing HPLC or ion chromatograph to easily determine anions in almost any sample matrix including: air, water and soil. They are well suited for use in systems employing suppressed/non-suppressed conductivity, electrochemical, UV, and ICP-MS detection.
Specifications
- Catalog No:
- HAMI79433
- HAMI79455
- Exchange capacity:
- 0,19 meq/gm
- 0,19 meq/gm
- Max. pressure:
- 5000 psi
- 5000 psi
- pH range:
- 1 - 13
- 1 - 13
- Temperature limit:
- pH 1 - 7,9 (5 - 60 °C); pH 8 - 13 (5 - 30 °C)
- pH 1 - 7,9 (5 - 60 °C); pH 8 - 13 (5 - 30 °C)