Order Entry
Singapore
ContactUsLinkComponent
Reversed phase HPLC columns, PRP-1
Reversed phase HPLC columns, PRP-1
Supplier:  HAMILTON BONADUZ
Certificates
undefined
Reversed phase HPLC columns, PRP-1
Supplier:  HAMILTON BONADUZ
Specifications
  • Catalog No:
  • HAMI79571
  • HAMI79381
  • HAMI79820
  • Max. pressure:
  • 5000 psi
  • 5000 psi
  • 5000 psi
  • pH range:
  • 1 - 13
  • 1 - 13
  • 1 - 13
  • USP code:
  • L21
  • L21
  • L21
Specifications