VWR®, Boxes for Broken Glass
Corrugated cardboard with PE pouch, 50 µm thick.
- Folded flat for space-saving storage
- A flap is built into the lid for easy filling
- The internal safety lid is closed for removal of the container
Ideal for collecting, transporting and removing broken glass from the laboratory. Consists of a sturdy box made of strong corrugated cardboard, and a PE insertion pouch which prevents any possible escape of fluids adhering to broken glass.
Does not qualify as a DOT container.
: Delivered unassembled. Assembly takes a few seconds.