Hippuric acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Benzoyl amidoacetic acid, Benzoylaminoacetic acid, Bz-Gly-OH, Benzoyl glycocoll, 2-Benzamidoacetic acid, N-Benzoylglycine
CAS Number:495-69-2
:
Formula:C₆H₅C(O)NHCH₂COOH
MDL Number:MFCD00002692
Boiling Pt:399
Melting Pt:188…191 °C
Density:1.371
Molecular Weight:179.18 g/mol
Merck Index:13,04735
EINECS:207-806-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: