Riboflavine (vitamin B2)
Catalog # EDQMR0600000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:130 mg
- Seq:2495
- Environmentally Preferable:
Specifications
About this item
Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G
:
:376.37 g/mol
Melting Pt:280
Storage Temperature:Ambient
EINECS:201-507-1
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022
Merck Index:12,08367