Calcium formate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Formic acid calcium salt
CAS Number:544-17-2
:
Formula:C₂H₂CaO₄
MDL Number:MFCD00036108
Density:2.02
Molecular Weight:130.11 g/mol
Melting Pt:>300 °C
Merck Index:12,01711
EINECS:208-863-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: