4-Bromobutyronitrile ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Bromobutanenitrile
CAS Number:5332-06-9
:
Formula:BrH₂CCH₂CH₂CN
MDL Number:MFCD00001971
Boiling Pt:204…206 °C
Density:1.491
Molecular Weight:148 g/mol
Flash Pt:103 °C (217 °F)
EINECS:226-236-6
UN:3276
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable: