di-Sodium succinate hexahydrate
: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate
:6106-21-4
:
:NaOOCCH₂CH₂COONa·6H₂O
:MFCD00149117
:1.529 g/cm³ (20 °C)
:270.14 g/mol
:120 °C (-6H₂O)
:13,08754
:205-778-7
:GHS07
:Ambient
- Environmentally Preferable: