Tetrakis(triphenylphosphine)palladium(0) (≥9% Pd) ≥99.8% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4
CAS Number:14221-01-3
:
Formula:C₇₂H₆₀P₄Pd
MDL Number:MFCD00010012
Density:1 g/cm³ (20 °C)
Molecular Weight:1155.58 g/mol
Melting Pt:100…105 °C
Storage Temperature:Freezer
EINECS:238-086-9
Specifications
- Environmentally Preferable: