Ladle, long handle, disposable Bio
: BURKLE
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container. A long handle with indentations for fingers ensures a firm hold and precise work.
- Product made from renewable raw materials
- Bio-PE, white
- Individually packaged and optionally sterilised, for single use
- Reaches into any corner
- Handle length 300 mm
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
The disposable ladle can be used in the pharmaceutical industry, chemistry and cosmetics and the food and animal feed industries.
- Catalog No:
- BURK5379-1026
- BURK5379-0026
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact, Sustainable Materials
- Environmental certification:
- ACT-Label
- ACT-Label