Order Entry
Singapore
ContactUsLinkComponent
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Catalog # VWRC301696S
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Catalog # VWRC301696S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (Active chlorine)(W/V)(at filling):
    Min. 12 %
  • Alkalinity:
    Max. 0.50 meq/g
  • Hg (Mercury):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :74.44 g/mol
EINECS:231-668-3
UN:1791
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120
Melting Pt:–30…–20 °C
Density:1.22 g/cm³ (20 °C)
ADR:8 II
Merck Index:13,08702