VWR® Gurr™ Histological Fixatives, AFA
VWRC81009.362
:
- Pack type:Plastic container
- Pk:5 L
- Description:Histological fixative, AFA
- Seq:03
- Environmentally Preferable:
Acidified alcoholic formalin for the fixation of glycogen.
Contains acetic acid, ethanol and formaldehyde.