Cell culture media, Minimum Essential Medium (MEM)
MEM is an advancement of the BME and the base medium of many further modifications. Because BME did not fulfil all requirements for some mammalian and HeLa cells, a better variation had to be developed. Today, MEM is one of the most used synthetic media and shows its versatility by supplementing with amino acids including Hank's or Earle's salts. Even the addition of only small amounts of FBS results in a positive effect on cell growth.
- Store from 2 to 8 °C
- Environmentally Preferable: