VWR®, Flash point reference standard, Abel Closed Cup, jet aviation fuel
VWRC99116.230
:
- Pack type:Aluminium bottle
- Pk:250 mL
- Description:Flash Point Reference Standard, Abel Closed Cup, Jet Aviation Fuel
- Environmentally Preferable:
- Flash point:41,2 °C
- Test method:IP 170, ISO 13736
Flash point Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Abel Closed Cup (IP 170, ISO 13736).
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Fully traceable to international standards
- Supplied with MSDS
- Tamper-evident security packaging
- 12 month shelf life
: UKAS ISO 17025 and ISO 17034.
: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.