Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
Boiling Pt:290
Molecular Weight:92.09 g/mol
Melting Pt:18.6
Flash Pt:177
Merck Index:13,04497
Storage Temperature:Ambient
EINECS:200-289-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:83.5 - 88.5%
- Appearance::Conforms
- Identification B (IR):Passes Test
- Appearance of Solution:Passes Test
- Acidity or Alkalinity:Passes Test
- Refractive index (20 °C):1449 - 1455
- Aldehydes:Max. 10 ppm
- Esters:Passes Test
- Impurity A and Related Substances:Passes Test
- Diethylene Glycol:Max. 0.1%
- Ethylene Glycol:Max. 0.1%
- Any Other Impurity (tRMax. 0.1%
- Individual Impurities:Max. 0.1%
- Total Impurities:Max. 1.0%
- Total of All Impurities (tR>Glycerol):Max. 0.5%
- Disregard Limit:Max. 0.05%
- Halogenated Compounds:Max. 30 ppm
- Sugars:Passes Test
- Cl (Chloride):Max. 10 ppm
- Heavy Metals (as Pb):Max. 5 ppm
- Water:12.0 - 16.0%
- Sulphated ash:Max. 0.01%
- Residual Solvents:Passes Test
- As (Arsenic):Max. 1 ppm
- Cd (Cadmium):V (Vanadium)
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 1 ppm
- Ni (Nickel):Max. 2 ppm
- Pb (Lead):Max. 2 ppm
- V (Vanadium):Max. 1 ppm
- Conforms to Ph. Eur.:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot NumberGlycerine 85% Ph. Eur.
Additional Product Details
spec_VWRC24384.290 spec_VWRC24384.290 spec_VWRC24384.320 spec_VWRC24384.320 spec_VWRC24384.460
Product specification
More
VWR Chemicals for Biopharma and Pharma Production